The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004947.4 | NP_004938.1 | 45 - 46 | 4789 | 69 |
XM_005264914.3 | XP_005264971.1 | 45 - 46 | 5061 | 69 | |
XM_005264915.3 | XP_005264972.1 | 45 - 46 | 5061 | 69 | |
XM_005264916.3 | XP_005264973.1 | 44 - 45 | 4812 | 69 | |
XM_005264917.3 | XP_005264974.1 | 45 - 46 | 5019 | 69 | |
XM_005264918.3 | XP_005264975.1 | 44 - 45 | 4691 | 69 | |
XM_006713008.3 | XP_006713071.1 | 45 - 46 | 5061 | 69 | |
XM_006713009.3 | XP_006713072.1 | 44 - 45 | 4974 | 69 | |
XM_006713010.3 | XP_006713073.1 | 45 - 46 | 4824 | 69 | |
XM_011533441.2 | XP_011531743.1 | 34 - 35 | 5670 | 69 | |
XM_011533443.2 | XP_011531745.1 | 21 - 22 | 3351 | 69 | |
XM_011533444.2 | XP_011531746.1 | 21 - 22 | 3275 | 69 | |
XM_011533445.2 | XP_011531747.1 | 21 - 22 | 3296 | 69 | |
XM_017005825.1 | XP_016861314.1 | 45 - 46 | 4840 | 69 | |
XM_017005826.1 | XP_016861315.1 | 45 - 46 | 4830 | 69 | |
XM_017005827.1 | XP_016861316.1 | 36 - 37 | 3981 | 69 | |
XM_017005828.1 | XP_016861317.1 | 20 - 21 | 2706 | 69 | |
GenBank mRNA | AB002297.1 | - | 40 - 41 | 4397 | 69 |
AY145303.1 | - | 45 - 46 | 4766 | 69 | |
AY254099.1 | - | 45 - 46 | 4789 | 69 | |
BC131746.1 | - | 10 - 11 | 1157 | 69 |