Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZSCAN29
Entrez Gene ID
146050
Gene Name
zinc finger and SCAN domain containing 29
Gene Aliases
ZNF690, Zfp690
Chromosome Location
Chr.15: 43358172 - 43371032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152455.3 NP_689668.3 3 - 4 1363 89
XM_006720401.3 XP_006720464.1 4 - 5 2123 89
XM_011521266.2 XP_011519568.1 4 - 5 1803 89
GenBank mRNA AF525399.1 - 3 - 4 1213 89
AK093186.1 - 2 - 3 909 89
AK093377.1 - 1 - 2 398 89
AK122788.1 - 3 - 4 1351 89
BC017179.2 - 1 - 2 229 89
BC040682.1 - 1 - 2 229 89
BC109270.1 - 3 - 4 1213 89

View Full Product Details