The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006358.3 | NP_006349.1 | 2 - 3 | 271 | 69 |
NR_104235.1 | 3 - 4 | 446 | 69 | ||
NR_104236.1 | 2 - 3 | 271 | 69 | ||
GenBank mRNA | AK292924.1 | - | 2 - 3 | 209 | 69 |
BC005957.1 | - | 2 - 3 | 209 | 69 | |
BC012998.2 | - | 2 - 3 | 191 | 69 | |
BU595416.1 | - | 2 - 3 | 264 | 69 | |
BX352732.2 | - | 2 - 3 | 126 | 69 | |
BX647991.1 | - | 2 - 3 | 203 | 69 | |
CR456577.1 | - | 2 - 3 | 187 | 69 | |
Y12860.1 | - | 2 - 3 | 241 | 69 |