The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291969.1 | NP_001278898.1 | 5 - 6 | 822 | 67 |
NM_001291970.1 | NP_001278899.1 | 7 - 8 | 1181 | 67 | |
NM_006502.2 | NP_006493.1 | 7 - 8 | 1181 | 67 | |
XM_005249186.3 | XP_005249243.1 | 6 - 7 | 1034 | 67 | |
XM_011514698.2 | XP_011513000.1 | 6 - 7 | 955 | 67 | |
GenBank mRNA | AB024313.1 | - | 7 - 8 | 1114 | 67 |
AF158185.1 | - | 7 - 8 | 1042 | 67 | |
AJ420484.1 | - | 4 - 5 | 498 | 67 | |
AK023893.1 | - | 7 - 8 | 1172 | 67 | |
AK025631.1 | - | - | 509 | 67 | |
AK122807.1 | - | 4 - 5 | 1089 | 67 | |
AK294430.1 | - | 7 - 8 | 1162 | 67 | |
AK307660.1 | - | 5 - 6 | 807 | 67 | |
AK316151.1 | - | 6 - 7 | 996 | 67 | |
BC015742.1 | - | 7 - 8 | 1025 | 67 |