The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291969.1 | NP_001278898.1 | 7 - 8 | 1019 | 102 |
NM_001291970.1 | NP_001278899.1 | 9 - 10 | 1378 | 102 | |
NM_006502.2 | NP_006493.1 | 9 - 10 | 1378 | 102 | |
XM_005249186.3 | XP_005249243.1 | 8 - 9 | 1231 | 102 | |
XM_011514698.2 | XP_011513000.1 | 8 - 9 | 1152 | 102 | |
GenBank mRNA | AB024313.1 | - | 9 - 10 | 1311 | 102 |
AF158185.1 | - | 9 - 10 | 1239 | 102 | |
AJ420484.1 | - | 6 - 7 | 695 | 102 | |
AK023893.1 | - | 9 - 10 | 1369 | 102 | |
AK025631.1 | - | - | 706 | 102 | |
AK122807.1 | - | 6 - 7 | 1286 | 102 | |
AK294430.1 | - | 9 - 10 | 1359 | 102 | |
AK307660.1 | - | 7 - 8 | 1004 | 102 | |
AK316151.1 | - | 8 - 9 | 1193 | 102 | |
BC015742.1 | - | 9 - 10 | 1222 | 102 |