The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015613.2 | NP_056428.1 | 2 - 3 | 614 | 94 |
XM_011539623.2 | XP_011537925.1 | 1 - 2 | 870 | 94 | |
GenBank mRNA | AB031547.1 | - | 2 - 3 | 614 | 94 |