The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020143.2 | NP_064528.1 | 5 - 6 | 678 | 102 |
GenBank mRNA | AF164799.1 | - | 5 - 6 | 636 | 102 |
AF349314.1 | - | 5 - 6 | 658 | 102 | |
AK223118.1 | - | 5 - 6 | 606 | 102 | |
AK225641.1 | - | 5 - 6 | 602 | 102 | |
AK291715.1 | - | 5 - 6 | 666 | 102 | |
AK313204.1 | - | 5 - 6 | 626 | 102 | |
BC008304.1 | - | 5 - 6 | 678 | 102 | |
BC018152.1 | - | 5 - 6 | 473 | 102 | |
L48692.1 | - | 5 - 6 | 613 | 102 |