Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NETO2
Entrez Gene ID
81831
Gene Name
neuropilin and tolloid like 2
Gene Aliases
BTCL2, NEOT2
Chromosome Location
Chr.16: 47081520 - 47144025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201477.1 NP_001188406.1 6 - 7 1037 91
NM_018092.4 NP_060562.3 6 - 7 1058 91
XM_006721289.1 XP_006721352.1 6 - 7 1058 91
XM_006721291.1 XP_006721354.1 6 - 7 1037 91
XM_006721292.1 XP_006721355.1 6 - 7 1058 91
XM_017023740.1 XP_016879229.1 6 - 7 772 91
GenBank mRNA AK074937.1 - 6 - 7 1030 91
AK297408.1 - 3 - 4 383 91
AL834404.1 - 3 - 4 424 91
AY358718.1 - 6 - 7 955 91
BC065504.1 - 6 - 7 739 91
BC108727.1 - 6 - 7 944 91

View Full Product Details