The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077500.1 | NP_001070968.1 | 3 - 4 | 389 | 136 |
NM_002776.4 | NP_002767.2 | 3 - 4 | 488 | 136 | |
NM_145888.2 | NP_665895.1 | 3 - 4 | 350 | 136 | |
XM_005259061.3 | XP_005259118.1 | 3 - 4 | 410 | 136 | |
XM_005259062.3 | XP_005259119.1 | 3 - 4 | 374 | 136 | |
XM_006723287.3 | XP_006723350.1 | 3 - 4 | 377 | 136 | |
XM_006723289.3 | XP_006723352.1 | 3 - 4 | 472 | 136 | |
XM_017026993.1 | XP_016882482.1 | 3 - 4 | 387 | 136 | |
GenBank mRNA | AK292365.1 | - | 3 - 4 | 479 | 136 |
AK309456.1 | - | 2 - 3 | 576 | 136 | |
AY561635.1 | - | 3 - 4 | 417 | 136 | |
BC002710.2 | - | 3 - 4 | 488 | 136 | |
BM830577.1 | - | 3 - 4 | 389 | 136 | |
CR450327.1 | - | 2 - 3 | 269 | 136 | |
KJ010190.1 | - | 2 - 3 | 202 | 136 | |
KJ010191.1 | - | 2 - 3 | 202 | 136 | |
KJ010197.1 | - | 2 - 3 | 202 | 136 | |
KJ010201.1 | - | 1 - 2 | 203 | 136 |