The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313915.1 | NP_001300844.1 | 16 - 17 | 3021 | 106 |
NM_004836.6 | NP_004827.4 | 16 - 17 | 3447 | 106 | |
XM_017005376.1 | XP_016860865.1 | 17 - 18 | 3339 | 106 | |
GenBank mRNA | AF110146.1 | - | 16 - 17 | 3214 | 106 |
AF193339.1 | - | 16 - 17 | 3444 | 106 | |
AK128659.1 | - | 17 - 18 | 3200 | 106 | |
AK308312.1 | - | 15 - 16 | 2813 | 106 | |
AK315287.1 | - | 16 - 17 | 3352 | 106 | |
BC126354.1 | - | 16 - 17 | 3241 | 106 | |
BC126356.1 | - | 16 - 17 | 3238 | 106 | |
CR749382.1 | - | 16 - 17 | 3022 | 106 |