The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313915.1 | NP_001300844.1 | 2 - 3 | 311 | 127 |
NM_004836.6 | NP_004827.4 | 2 - 3 | 737 | 127 | |
XM_017005376.1 | XP_016860865.1 | 2 - 3 | 433 | 127 | |
GenBank mRNA | AF110146.1 | - | 2 - 3 | 504 | 127 |
AF193339.1 | - | 2 - 3 | 734 | 127 | |
AK128659.1 | - | 3 - 4 | 490 | 127 | |
AK315287.1 | - | 2 - 3 | 642 | 127 | |
BC126354.1 | - | 2 - 3 | 531 | 127 | |
BC126356.1 | - | 2 - 3 | 528 | 127 | |
CR749382.1 | - | 2 - 3 | 312 | 127 |