The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164232.1 | NP_001157704.1 | 3 - 4 | 602 | 68 |
NM_001164234.1 | NP_001157706.1 | 3 - 4 | 779 | 68 | |
NM_015214.2 | NP_056029.2 | 3 - 4 | 779 | 68 | |
XM_005273454.2 | XP_005273511.1 | 3 - 4 | 927 | 68 | |
XM_005273455.3 | XP_005273512.1 | 3 - 4 | 927 | 68 | |
XM_005273456.3 | XP_005273513.1 | 3 - 4 | 927 | 68 | |
XM_011544456.1 | XP_011542758.1 | 3 - 4 | 927 | 68 | |
XM_017013252.1 | XP_016868741.1 | 3 - 4 | 736 | 68 | |
XM_017013253.1 | XP_016868742.1 | 3 - 4 | 927 | 68 | |
XM_017013254.1 | XP_016868743.1 | 3 - 4 | 584 | 68 | |
XM_017013255.1 | XP_016868744.1 | 3 - 4 | 683 | 68 | |
XM_017013256.1 | XP_016868745.1 | 3 - 4 | 683 | 68 | |
GenBank mRNA | AK125904.1 | - | 3 - 4 | 602 | 68 |
AK299875.1 | - | 2 - 3 | 398 | 68 |