Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC6A4
Entrez Gene ID
6532
Gene Name
solute carrier family 6 member 4
Gene Aliases
5-HTT, 5-HTTLPR, 5HTT, HTT, OCD1, SERT, SERT1, hSERT
Chromosome Location
Chr.17: 30194319 - 30235968 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045.5 NP_001036.1 - 916 98
GenBank mRNA AK308014.1 - 3 - 4 647 98
AK313166.1 - 3 - 4 645 98
AY902473.1 - 1 - 2 411 98
BC069484.1 - 1 - 2 388 98
BX399758.2 - 3 - 4 675 98
L05568.1 - 1 - 2 412 98
X70697.1 - 1 - 2 450 98

View Full Product Details