The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012514.2 | NP_001012532.1 | 4 - 5 | 705 | 90 |
NM_001012516.2 | NP_001012534.1 | 4 - 5 | 680 | 90 | |
NM_001287240.1 | NP_001274169.1 | 5 - 6 | 782 | 90 | |
NM_001287241.1 | NP_001274170.1 | 6 - 7 | 978 | 90 | |
NM_030926.5 | NP_112188.1 | 5 - 6 | 846 | 90 | |
GenBank mRNA | AB003629.1 | - | 5 - 6 | 715 | 90 |
AF271781.1 | - | 5 - 6 | 831 | 90 | |
AF272043.1 | - | 5 - 6 | 828 | 90 | |
AK056321.1 | - | 4 - 5 | 652 | 90 | |
AK075361.1 | - | 5 - 6 | 811 | 90 | |
AK090975.1 | - | 5 - 6 | 757 | 90 | |
AK222697.1 | - | 5 - 6 | 793 | 90 | |
AK295503.1 | - | 5 - 6 | 612 | 90 | |
AK303199.1 | - | 5 - 6 | 800 | 90 | |
AK307702.1 | - | 4 - 5 | 614 | 90 | |
AL136603.1 | - | 5 - 6 | 787 | 90 | |
AL713651.1 | - | 5 - 6 | 770 | 90 | |
AY049777.1 | - | 5 - 6 | 732 | 90 | |
BC002424.1 | - | 5 - 6 | 773 | 90 | |
BC025742.1 | - | 4 - 5 | 650 | 90 | |
BC050668.1 | - | 4 - 5 | 640 | 90 | |
BC098563.1 | - | 5 - 6 | 759 | 90 | |
CR457208.1 | - | 5 - 6 | 715 | 90 |