The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282541.1 | NP_001269470.1 | 30 - 31 | 2946 | 200 |
NM_001282542.1 | NP_001269471.1 | 29 - 30 | 2871 | 200 | |
NM_003176.3 | NP_003167.2 | 30 - 31 | 2943 | 200 | |
XM_005271155.3 | XP_005271212.1 | 29 - 30 | 3267 | 200 | |
XM_005271156.1 | XP_005271213.1 | 29 - 30 | 2741 | 200 | |
XM_006710859.1 | XP_006710922.1 | 30 - 31 | 2854 | 200 | |
XM_011542038.2 | XP_011540340.1 | 23 - 24 | 2137 | 200 | |
XM_017002184.1 | XP_016857673.1 | 30 - 31 | 2746 | 200 | |
XM_017002185.1 | XP_016857674.1 | 24 - 25 | 2172 | 200 | |
GenBank mRNA | AK315268.1 | - | 29 - 30 | 2830 | 200 |
BC126266.1 | - | 30 - 31 | 2820 | 200 | |
BC144019.1 | - | 30 - 31 | 2823 | 200 | |
D67035.1 | - | 30 - 31 | 2870 | 200 | |
X95654.1 | - | 30 - 31 | 2797 | 200 |