Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERGEF
Entrez Gene ID
26297
Gene Name
secretion regulating guanine nucleotide exchange factor
Gene Aliases
DELGEF, Gnefr
Chromosome Location
Chr.11: 17788048 - 18013162 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012139.3 NP_036271.1 7 - 8 843 65
NR_104040.1 7 - 8 843 65
NR_104041.1 7 - 8 843 65
GenBank mRNA AJ243950.1 - 7 - 8 709 65
AJ243951.1 - 7 - 8 709 65
AK292286.1 - 7 - 8 843 65
AK294022.1 - 7 - 8 708 65
BC000707.2 - 7 - 8 717 65
BC065375.1 - 7 - 8 823 65
BE075522.1 - 4 - 5 289 65

View Full Product Details