Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDHD2
Entrez Gene ID
23259
Gene Name
DDHD domain containing 2
Gene Aliases
SAMWD1, SPG54, iPLA(1)gamma
Chromosome Location
Chr.8: 38231343 - 38273442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005273454.2 XP_005273511.1 17 - 18 2681 99
XM_017013256.1 XP_016868745.1 16 - 17 2369 99

View Full Product Details