Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL21A1
Entrez Gene ID
81578
Gene Name
collagen type XXI alpha 1 chain
Gene Aliases
COLA1L, FP633
Chromosome Location
Chr.6: 56056590 - 56394128 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318751.1 NP_001305680.1 17 - 18 2063 99
NM_001318752.1 NP_001305681.1 15 - 16 1944 99
NM_030820.3 NP_110447.2 16 - 17 1989 99
NR_134849.1 16 - 17 1981 99
XM_006715223.1 XP_006715286.1 16 - 17 2155 99
XM_011514924.2 XP_011513226.1 16 - 17 1932 99
XM_011514925.2 XP_011513227.1 18 - 19 2102 99
XM_011514926.1 XP_011513228.1 16 - 17 1860 99
XM_011514927.1 XP_011513229.1 16 - 17 1814 99
GenBank mRNA AF330693.1 - 15 - 16 1910 99
AF370383.1 - 3 - 4 149 99
AF414088.1 - 16 - 17 1959 99
AK313398.1 - 16 - 17 1965 99
AL136624.1 - 16 - 17 1959 99
BC045597.1 - 16 - 17 1936 99
BC126108.1 - 16 - 17 1844 99
BC143862.1 - 16 - 17 1836 99
BC143865.1 - 17 - 18 1918 99

View Full Product Details