The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003632.2 | NP_003623.1 | 16 - 17 | 2743 | 64 |
XM_005257748.4 | XP_005257805.1 | 16 - 17 | 3508 | 64 | |
XM_017025238.1 | XP_016880727.1 | 18 - 19 | 3878 | 64 | |
GenBank mRNA | U87223.1 | - | 16 - 17 | 2744 | 64 |