The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003632.2 | NP_003623.1 | 1 - 2 | 286 | 85 |
XM_017025238.1 | XP_016880727.1 | 3 - 4 | 1421 | 85 | |
GenBank mRNA | U87223.1 | - | 1 - 2 | 287 | 85 |