The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003632.2 | NP_003623.1 | 7 - 8 | 1262 | 55 |
XM_005257748.4 | XP_005257805.1 | 7 - 8 | 2027 | 55 | |
XM_017025238.1 | XP_016880727.1 | 9 - 10 | 2397 | 55 | |
GenBank mRNA | U87223.1 | - | 7 - 8 | 1263 | 55 |