The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004719.2 | NP_004710.2 | 2 - 3 | 341 | 151 |
XM_017020219.1 | XP_016875708.1 | 2 - 3 | 388 | 151 | |
XM_017020223.1 | XP_016875712.1 | 2 - 3 | 388 | 151 | |
GenBank mRNA | AF030234.1 | - | 2 - 3 | 288 | 151 |
AK302999.1 | - | 2 - 3 | 266 | 151 | |
BC017851.1 | - | 2 - 3 | 149 | 151 |