The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247996.1 | NP_001234925.1 | 17 - 18 | 1715 | 73 |
NM_018482.3 | NP_060952.2 | 16 - 17 | 1613 | 73 | |
XM_005250925.1 | XP_005250982.1 | 17 - 18 | 1622 | 73 | |
XM_006716563.3 | XP_006716626.1 | 17 - 18 | 1441 | 73 | |
XM_006716564.1 | XP_006716627.1 | 18 - 19 | 1600 | 73 | |
XM_006716565.3 | XP_006716628.1 | 16 - 17 | 1437 | 73 | |
XM_006716566.1 | XP_006716629.1 | 17 - 18 | 1622 | 73 | |
XM_011517052.2 | XP_011515354.1 | 17 - 18 | 1438 | 73 | |
XM_011517053.1 | XP_011515355.1 | 17 - 18 | 1644 | 73 | |
XM_017013467.1 | XP_016868956.1 | 15 - 16 | 1459 | 73 | |
XM_017013468.1 | XP_016868957.1 | 16 - 17 | 1683 | 73 | |
XM_017013469.1 | XP_016868958.1 | 16 - 17 | 1613 | 73 | |
GenBank mRNA | AB033075.1 | - | 10 - 11 | 846 | 73 |
BC137135.1 | - | 15 - 16 | 1410 | 73 | |
BC144493.1 | - | 16 - 17 | 1512 | 73 | |
BX537768.1 | - | 9 - 10 | 866 | 73 |