The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193520.1 | NP_001180449.1 | 18 - 19 | 2271 | 54 |
NM_001193521.1 | NP_001180450.1 | 17 - 18 | 2172 | 54 | |
NM_001301202.1 | NP_001288131.1 | 17 - 18 | 2104 | 54 | |
NM_004658.2 | NP_004649.2 | 18 - 19 | 2280 | 54 | |
XM_005253950.4 | XP_005254007.1 | 18 - 19 | 2286 | 54 | |
XM_006719641.3 | XP_006719704.1 | 18 - 19 | 2125 | 54 | |
XM_006719642.3 | XP_006719705.1 | 17 - 18 | 2255 | 54 | |
XM_011538852.2 | XP_011537154.1 | 17 - 18 | 2258 | 54 | |
XM_011538853.2 | XP_011537155.1 | 17 - 18 | 2252 | 54 | |
XM_011538854.2 | XP_011537156.1 | 7 - 8 | 998 | 54 | |
XM_017020028.1 | XP_016875517.1 | 18 - 19 | 2283 | 54 | |
XM_017020029.1 | XP_016875518.1 | 17 - 18 | 2258 | 54 | |
XM_017020030.1 | XP_016875519.1 | 18 - 19 | 2286 | 54 | |
XM_017020031.1 | XP_016875520.1 | 18 - 19 | 2283 | 54 | |
GenBank mRNA | AB208935.1 | - | 18 - 19 | 2256 | 54 |
AF086713.1 | - | 18 - 19 | 2257 | 54 | |
AK294350.1 | - | 18 - 19 | 2283 | 54 | |
AL136672.1 | - | 17 - 18 | 2199 | 54 | |
BC014420.1 | - | 17 - 18 | 2063 | 54 | |
BC093724.1 | - | 17 - 18 | 2006 | 54 | |
BC143261.1 | - | 17 - 18 | 2009 | 54 |