Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAST
Entrez Gene ID
831
Gene Name
calpastatin
Gene Aliases
BS-17, PLACK
Chromosome Location
Chr.5: 96662037 - 96774683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042440.3 NP_001035905.1 16 - 17 1476 125
NM_001190442.1 NP_001177371.1 17 - 18 1348 125
NM_001284212.2 NP_001271141.1 14 - 15 1127 125
NM_001284213.2 NP_001271142.1 13 - 14 1034 125
NM_173060.3 NP_775083.1 15 - 16 1184 125
NR_104285.1 3 - 4 340 125
XM_006714696.3 XP_006714759.1 19 - 20 1490 125
XM_006714697.3 XP_006714760.1 18 - 19 1507 125
XM_006714698.3 XP_006714761.1 17 - 18 1468 125
XM_006714699.3 XP_006714762.1 18 - 19 1431 125
XM_006714700.3 XP_006714763.1 17 - 18 1450 125
XM_006714701.3 XP_006714764.1 17 - 18 1466 125
XM_006714702.3 XP_006714765.1 17 - 18 1342 125
XM_006714703.3 XP_006714766.1 16 - 17 1427 125
XM_006714704.3 XP_006714767.1 17 - 18 1375 125
XM_006714705.3 XP_006714768.1 16 - 17 1384 125
XM_006714706.3 XP_006714769.1 16 - 17 1302 125
XM_006714707.3 XP_006714770.1 16 - 17 1253 125
XM_006714708.3 XP_006714771.1 15 - 16 1213 125
XM_006714709.3 XP_006714772.1 15 - 16 1176 125
XM_006714710.3 XP_006714773.1 14 - 15 1131 125
XM_006714711.3 XP_006714774.1 14 - 15 1176 125
XM_006714712.3 XP_006714775.1 13 - 14 1123 125
XM_006714713.3 XP_006714776.1 3 - 4 340 125
XM_006714714.3 XP_006714777.1 3 - 4 340 125
XM_006714715.3 XP_006714778.1 4 - 5 1066 125
XM_011543654.2 XP_011541956.1 17 - 18 1426 125
XM_011543655.2 XP_011541957.1 17 - 18 1423 125
XM_011543656.2 XP_011541958.1 16 - 17 1370 125
XM_011543657.2 XP_011541959.1 16 - 17 1252 125
XM_011543658.2 XP_011541960.1 14 - 15 1105 125
XM_017009911.1 XP_016865400.1 17 - 18 1468 125
XM_017009912.1 XP_016865401.1 18 - 19 1479 125
XM_017009913.1 XP_016865402.1 18 - 19 1448 125
XM_017009914.1 XP_016865403.1 17 - 18 1450 125
XM_017009915.1 XP_016865404.1 18 - 19 1452 125
XM_017009916.1 XP_016865405.1 17 - 18 1466 125
XM_017009917.1 XP_016865406.1 17 - 18 1384 125
XM_017009918.1 XP_016865407.1 16 - 17 1411 125
XM_017009919.1 XP_016865408.1 16 - 17 1384 125
XM_017009920.1 XP_016865409.1 16 - 17 1294 125
XM_017009921.1 XP_016865410.1 16 - 17 1311 125
XM_017009922.1 XP_016865411.1 15 - 16 1345 125
XM_017009923.1 XP_016865412.1 15 - 16 1345 125
XM_017009924.1 XP_016865413.1 15 - 16 1327 125
XM_017009925.1 XP_016865414.1 15 - 16 1263 125
XM_017009926.1 XP_016865415.1 15 - 16 1263 125
XM_017009927.1 XP_016865416.1 16 - 17 1252 125
XM_017009928.1 XP_016865417.1 15 - 16 1251 125
XM_017009929.1 XP_016865418.1 15 - 16 1170 125
XM_017009930.1 XP_016865419.1 14 - 15 1137 125
XM_017009931.1 XP_016865420.1 14 - 15 1131 125
XM_017009932.1 XP_016865421.1 13 - 14 1194 125
GenBank mRNA AB208816.1 - 17 - 18 1312 125
AF327443.1 - 12 - 13 1026 125
AK292289.1 - 3 - 4 340 125
AK292356.1 - 3 - 4 313 125
AK296919.1 - 17 - 18 1306 125
AK297879.1 - 15 - 16 1126 125
AK298541.1 - 16 - 17 1253 125
AK299402.1 - 16 - 17 1233 125
AK300624.1 - 13 - 14 1017 125
AK302206.1 - 4 - 5 340 125
AK311585.1 - 4 - 5 518 125
AK311598.1 - 4 - 5 394 125
AK314290.1 - 14 - 15 1129 125
AK315146.1 - 14 - 15 1128 125
AK315673.1 - 3 - 4 288 125
AL832174.1 - 15 - 16 1169 125
AL832349.1 - 15 - 16 1172 125
AY189741.1 - 5 - 6 339 125
BC013579.1 - 16 - 17 1179 125
BC078149.1 - 11 - 12 762 125
BT009783.1 - 13 - 14 961 125
BX648397.1 - 14 - 15 1078 125
D16217.1 - 16 - 17 1246 125
DB033043.1 - 3 - 4 288 125
U26724.2 - 7 - 8 609 125
U31345.1 - 5 - 6 356 125
U58996.2 - 3 - 4 319 125

View Full Product Details