Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAST
Entrez Gene ID
831
Gene Name
calpastatin
Gene Aliases
BS-17, PLACK
Chromosome Location
Chr.5: 96662037 - 96774683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042440.3 NP_001035905.1 25 - 26 2273 87
NM_001190442.1 NP_001177371.1 26 - 27 2145 87
NM_001284212.2 NP_001271141.1 23 - 24 1921 87
NM_001284213.2 NP_001271142.1 22 - 23 1831 87
NM_173060.3 NP_775083.1 24 - 25 1981 87
NR_104285.1 12 - 13 1134 87
XM_006714696.3 XP_006714759.1 28 - 29 2287 87
XM_006714697.3 XP_006714760.1 27 - 28 2301 87
XM_006714698.3 XP_006714761.1 26 - 27 2265 87
XM_006714699.3 XP_006714762.1 27 - 28 2228 87
XM_006714700.3 XP_006714763.1 26 - 27 2247 87
XM_006714701.3 XP_006714764.1 26 - 27 2263 87
XM_006714702.3 XP_006714765.1 26 - 27 2139 87
XM_006714703.3 XP_006714766.1 25 - 26 2224 87
XM_006714704.3 XP_006714767.1 26 - 27 2172 87
XM_006714705.3 XP_006714768.1 25 - 26 2178 87
XM_006714706.3 XP_006714769.1 25 - 26 2099 87
XM_006714707.3 XP_006714770.1 25 - 26 2050 87
XM_006714708.3 XP_006714771.1 24 - 25 2010 87
XM_006714709.3 XP_006714772.1 24 - 25 1973 87
XM_006714710.3 XP_006714773.1 23 - 24 1928 87
XM_006714711.3 XP_006714774.1 23 - 24 1973 87
XM_006714712.3 XP_006714775.1 22 - 23 1920 87
XM_006714713.3 XP_006714776.1 12 - 13 1137 87
XM_006714714.3 XP_006714777.1 12 - 13 1134 87
XM_006714715.3 XP_006714778.1 13 - 14 1863 87
XM_011543654.2 XP_011541956.1 26 - 27 2223 87
XM_011543655.2 XP_011541957.1 26 - 27 2220 87
XM_011543656.2 XP_011541958.1 25 - 26 2167 87
XM_011543657.2 XP_011541959.1 25 - 26 2049 87
XM_011543658.2 XP_011541960.1 23 - 24 1902 87
XM_017009911.1 XP_016865400.1 26 - 27 2262 87
XM_017009912.1 XP_016865401.1 27 - 28 2273 87
XM_017009913.1 XP_016865402.1 27 - 28 2245 87
XM_017009914.1 XP_016865403.1 26 - 27 2244 87
XM_017009915.1 XP_016865404.1 27 - 28 2249 87
XM_017009916.1 XP_016865405.1 26 - 27 2260 87
XM_017009917.1 XP_016865406.1 26 - 27 2181 87
XM_017009918.1 XP_016865407.1 25 - 26 2208 87
XM_017009919.1 XP_016865408.1 25 - 26 2181 87
XM_017009920.1 XP_016865409.1 25 - 26 2091 87
XM_017009921.1 XP_016865410.1 25 - 26 2105 87
XM_017009922.1 XP_016865411.1 24 - 25 2142 87
XM_017009923.1 XP_016865412.1 24 - 25 2139 87
XM_017009924.1 XP_016865413.1 24 - 25 2121 87
XM_017009925.1 XP_016865414.1 24 - 25 2060 87
XM_017009926.1 XP_016865415.1 24 - 25 2057 87
XM_017009927.1 XP_016865416.1 25 - 26 2046 87
XM_017009928.1 XP_016865417.1 24 - 25 2045 87
XM_017009929.1 XP_016865418.1 24 - 25 1964 87
XM_017009930.1 XP_016865419.1 23 - 24 1934 87
XM_017009931.1 XP_016865420.1 23 - 24 1925 87
XM_017009932.1 XP_016865421.1 22 - 23 1988 87
GenBank mRNA AB208816.1 - 26 - 27 2109 87
AF327443.1 - 21 - 22 1823 87
AK292289.1 - 12 - 13 1137 87
AK292356.1 - 12 - 13 1110 87
AK296919.1 - 26 - 27 2103 87
AK297879.1 - 24 - 25 1923 87
AK298541.1 - 25 - 26 2050 87
AK299384.1 - 23 - 24 1681 87
AK299402.1 - 25 - 26 2030 87
AK300624.1 - 22 - 23 1814 87
AK302206.1 - 13 - 14 1137 87
AK314290.1 - 23 - 24 1926 87
AK315146.1 - 23 - 24 1925 87
AK315673.1 - 12 - 13 1082 87
AL045308.1 - 5 - 6 413 87
AL832174.1 - 24 - 25 1966 87
AL832349.1 - 24 - 25 1969 87
AY189741.1 - 14 - 15 1136 87
BC013579.1 - 25 - 26 1976 87
BC078149.1 - 20 - 21 1535 87
BT009783.1 - 22 - 23 1758 87
BX648397.1 - 23 - 24 87
D16217.1 - 25 - 26 2043 87
D50827.1 - 3 - 4 291 87
S73329.1 - 4 - 5 315 87
U26724.2 - 16 - 17 1406 87
U31345.1 - 14 - 15 1150 87
U31346.1 - 4 - 5 366 87
U38525.1 - 7 - 8 609 87
U58996.2 - 12 - 13 1113 87

View Full Product Details