Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAST
Entrez Gene ID
831
Gene Name
calpastatin
Gene Aliases
BS-17, PLACK
Chromosome Location
Chr.5: 96662037 - 96774683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190442.1 NP_001177371.1 6 - 7 431 78
NM_001284213.2 NP_001271142.1 3 - 4 228 78
NM_173060.3 NP_775083.1 3 - 4 228 78
XM_006714696.3 XP_006714759.1 7 - 8 534 78
XM_006714697.3 XP_006714760.1 6 - 7 551 78
XM_006714698.3 XP_006714761.1 6 - 7 551 78
XM_006714699.3 XP_006714762.1 6 - 7 475 78
XM_006714701.3 XP_006714764.1 5 - 6 510 78
XM_006714703.3 XP_006714766.1 5 - 6 510 78
XM_006714704.3 XP_006714767.1 5 - 6 419 78
XM_006714707.3 XP_006714770.1 4 - 5 297 78
XM_006714708.3 XP_006714771.1 4 - 5 296 78
XM_006714710.3 XP_006714773.1 3 - 4 214 78
XM_011543654.2 XP_011541956.1 6 - 7 551 78
XM_011543655.2 XP_011541957.1 6 - 7 551 78
XM_011543656.2 XP_011541958.1 5 - 6 495 78
XM_011543657.2 XP_011541959.1 5 - 6 377 78
XM_011543658.2 XP_011541960.1 3 - 4 230 78
XM_017009911.1 XP_016865400.1 6 - 7 551 78
XM_017009912.1 XP_016865401.1 7 - 8 562 78
XM_017009915.1 XP_016865404.1 6 - 7 496 78
XM_017009916.1 XP_016865405.1 5 - 6 510 78
XM_017009917.1 XP_016865406.1 6 - 7 467 78
XM_017009919.1 XP_016865408.1 6 - 7 551 78
XM_017009920.1 XP_016865409.1 5 - 6 377 78
XM_017009921.1 XP_016865410.1 5 - 6 394 78
XM_017009927.1 XP_016865416.1 4 - 5 296 78
XM_017009928.1 XP_016865417.1 4 - 5 334 78
XM_017009929.1 XP_016865418.1 3 - 4 214 78
XM_017009931.1 XP_016865420.1 3 - 4 214 78
GenBank mRNA AB026049.1 - 4 - 5 283 78
AB208816.1 - 6 - 7 395 78
AK296919.1 - 5 - 6 350 78
AK297879.1 - 4 - 5 209 78
AK298541.1 - 4 - 5 297 78
AK299384.1 - 4 - 5 201 78
AK299402.1 - 4 - 5 277 78
AK300624.1 - 3 - 4 211 78
AK303900.1 - 3 - 4 215 78
AK314290.1 - 3 - 4 212 78
AL832174.1 - 3 - 4 213 78
AL832349.1 - 3 - 4 216 78
BC016066.1 - 5 - 6 531 78
BQ420953.1 - 4 - 5 277 78
CD702474.1 - 4 - 5 328 78
D16217.1 - 4 - 5 290 78
DC424542.1 - 3 - 4 228 78
DR004967.1 - 5 - 6 503 78
M86258.1 - 3 - 4 128 78

View Full Product Details