Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAST
Entrez Gene ID
831
Gene Name
calpastatin
Gene Aliases
BS-17, PLACK
Chromosome Location
Chr.5: 96662037 - 96774683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042440.3 NP_001035905.1 8 - 9 847 107
NM_001190442.1 NP_001177371.1 10 - 11 758 107
NM_001284212.2 NP_001271141.1 6 - 7 498 107
NM_173060.3 NP_775083.1 7 - 8 555 107
XM_006714696.3 XP_006714759.1 11 - 12 861 107
XM_006714697.3 XP_006714760.1 10 - 11 878 107
XM_006714698.3 XP_006714761.1 10 - 11 878 107
XM_006714699.3 XP_006714762.1 10 - 11 802 107
XM_006714700.3 XP_006714763.1 9 - 10 821 107
XM_006714701.3 XP_006714764.1 9 - 10 837 107
XM_006714702.3 XP_006714765.1 9 - 10 713 107
XM_006714703.3 XP_006714766.1 9 - 10 837 107
XM_006714704.3 XP_006714767.1 9 - 10 746 107
XM_006714705.3 XP_006714768.1 8 - 9 755 107
XM_006714706.3 XP_006714769.1 8 - 9 673 107
XM_006714707.3 XP_006714770.1 8 - 9 624 107
XM_006714708.3 XP_006714771.1 8 - 9 623 107
XM_006714709.3 XP_006714772.1 7 - 8 547 107
XM_006714710.3 XP_006714773.1 7 - 8 541 107
XM_006714711.3 XP_006714774.1 6 - 7 547 107
XM_006714712.3 XP_006714775.1 6 - 7 533 107
XM_011543654.2 XP_011541956.1 9 - 10 797 107
XM_011543655.2 XP_011541957.1 10 - 11 878 107
XM_011543656.2 XP_011541958.1 8 - 9 741 107
XM_011543657.2 XP_011541959.1 8 - 9 623 107
XM_011543658.2 XP_011541960.1 6 - 7 476 107
XM_017009911.1 XP_016865400.1 10 - 11 878 107
XM_017009912.1 XP_016865401.1 11 - 12 889 107
XM_017009913.1 XP_016865402.1 10 - 11 819 107
XM_017009914.1 XP_016865403.1 9 - 10 821 107
XM_017009915.1 XP_016865404.1 10 - 11 823 107
XM_017009916.1 XP_016865405.1 9 - 10 837 107
XM_017009917.1 XP_016865406.1 10 - 11 794 107
XM_017009918.1 XP_016865407.1 9 - 10 821 107
XM_017009919.1 XP_016865408.1 10 - 11 878 107
XM_017009920.1 XP_016865409.1 9 - 10 704 107
XM_017009921.1 XP_016865410.1 9 - 10 721 107
XM_017009922.1 XP_016865411.1 8 - 9 755 107
XM_017009923.1 XP_016865412.1 8 - 9 755 107
XM_017009924.1 XP_016865413.1 9 - 10 821 107
XM_017009925.1 XP_016865414.1 8 - 9 673 107
XM_017009926.1 XP_016865415.1 8 - 9 673 107
XM_017009927.1 XP_016865416.1 8 - 9 623 107
XM_017009928.1 XP_016865417.1 8 - 9 661 107
XM_017009929.1 XP_016865418.1 7 - 8 541 107
XM_017009930.1 XP_016865419.1 7 - 8 547 107
XM_017009931.1 XP_016865420.1 7 - 8 541 107
XM_017009932.1 XP_016865421.1 6 - 7 604 107
GenBank mRNA AB208816.1 - 10 - 11 722 107
AF327443.1 - 5 - 6 436 107
AK296919.1 - 9 - 10 677 107
AK297879.1 - 8 - 9 536 107
AK298541.1 - 8 - 9 624 107
AK299384.1 - 8 - 9 528 107
AK299402.1 - 8 - 9 604 107
AK303900.1 - 6 - 7 461 107
AK314290.1 - 7 - 8 539 107
AK315146.1 - 7 - 8 538 107
AL832174.1 - 7 - 8 540 107
AL832349.1 - 7 - 8 543 107
BC013579.1 - 8 - 9 550 107
BC016066.1 - 9 - 10 858 107
BC078149.1 - 3 - 4 229 107
BQ420953.1 - 7 - 8 523 107
BX648397.1 - 6 - 7 449 107
D16217.1 - 8 - 9 617 107
M33328.1 - 1 - 2 83 107
M86258.1 - 7 - 8 455 107

View Full Product Details