Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAST
Entrez Gene ID
831
Gene Name
calpastatin
Gene Aliases
BS-17, PLACK
Chromosome Location
Chr.5: 96662037 - 96774683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042440.3 NP_001035905.1 14 - 15 1341 78
NM_001190442.1 NP_001177371.1 15 - 16 1213 78
NM_001284212.2 NP_001271141.1 12 - 13 992 78
NM_001284213.2 NP_001271142.1 11 - 12 899 78
NM_173060.3 NP_775083.1 13 - 14 1049 78
XM_006714696.3 XP_006714759.1 17 - 18 1355 78
XM_006714697.3 XP_006714760.1 16 - 17 1372 78
XM_006714698.3 XP_006714761.1 15 - 16 1333 78
XM_006714699.3 XP_006714762.1 16 - 17 1296 78
XM_006714700.3 XP_006714763.1 15 - 16 1315 78
XM_006714701.3 XP_006714764.1 15 - 16 1331 78
XM_006714702.3 XP_006714765.1 15 - 16 1207 78
XM_006714703.3 XP_006714766.1 14 - 15 1292 78
XM_006714704.3 XP_006714767.1 15 - 16 1240 78
XM_006714705.3 XP_006714768.1 14 - 15 1249 78
XM_006714706.3 XP_006714769.1 14 - 15 1167 78
XM_006714707.3 XP_006714770.1 14 - 15 1118 78
XM_006714708.3 XP_006714771.1 13 - 14 1078 78
XM_006714709.3 XP_006714772.1 13 - 14 1041 78
XM_006714710.3 XP_006714773.1 12 - 13 996 78
XM_006714711.3 XP_006714774.1 12 - 13 1041 78
XM_006714712.3 XP_006714775.1 11 - 12 988 78
XM_006714715.3 XP_006714778.1 2 - 3 931 78
XM_011543654.2 XP_011541956.1 15 - 16 1291 78
XM_011543656.2 XP_011541958.1 14 - 15 1235 78
XM_011543657.2 XP_011541959.1 14 - 15 1117 78
XM_011543658.2 XP_011541960.1 12 - 13 970 78
XM_017009911.1 XP_016865400.1 15 - 16 1333 78
XM_017009912.1 XP_016865401.1 16 - 17 1344 78
XM_017009913.1 XP_016865402.1 16 - 17 1313 78
XM_017009914.1 XP_016865403.1 15 - 16 1315 78
XM_017009915.1 XP_016865404.1 16 - 17 1317 78
XM_017009916.1 XP_016865405.1 15 - 16 1331 78
XM_017009917.1 XP_016865406.1 15 - 16 1249 78
XM_017009918.1 XP_016865407.1 14 - 15 1276 78
XM_017009920.1 XP_016865409.1 14 - 15 1159 78
XM_017009921.1 XP_016865410.1 14 - 15 1176 78
XM_017009922.1 XP_016865411.1 13 - 14 1210 78
XM_017009923.1 XP_016865412.1 13 - 14 1210 78
XM_017009925.1 XP_016865414.1 13 - 14 1128 78
XM_017009926.1 XP_016865415.1 13 - 14 1128 78
XM_017009927.1 XP_016865416.1 14 - 15 1117 78
XM_017009928.1 XP_016865417.1 13 - 14 1116 78
XM_017009929.1 XP_016865418.1 13 - 14 1035 78
XM_017009930.1 XP_016865419.1 12 - 13 1002 78
XM_017009931.1 XP_016865420.1 12 - 13 996 78
XM_017009932.1 XP_016865421.1 11 - 12 1059 78
GenBank mRNA AB208816.1 - 15 - 16 1177 78
AF327443.1 - 10 - 11 891 78
AK296919.1 - 15 - 16 1171 78
AK297879.1 - 13 - 14 991 78
AK298541.1 - 14 - 15 1118 78
AK299384.1 - 14 - 15 1022 78
AK299402.1 - 14 - 15 1098 78
AK300624.1 - 11 - 12 882 78
AK302206.1 - 2 - 3 205 78
AK303836.1 - 7 - 8 572 78
AK303900.1 - 10 - 11 785 78
AK311585.1 - 2 - 3 383 78
AK311598.1 - 2 - 3 259 78
AK314290.1 - 12 - 13 994 78
AK315146.1 - 12 - 13 993 78
AL832174.1 - 13 - 14 1034 78
AL832349.1 - 13 - 14 1037 78
AY189741.1 - 3 - 4 204 78
BC013579.1 - 14 - 15 1044 78
BC078149.1 - 9 - 10 627 78
BT009783.1 - 11 - 12 826 78
BX648397.1 - 12 - 13 943 78
D16217.1 - 14 - 15 1111 78
U26724.2 - 5 - 6 474 78
U31345.1 - 3 - 4 221 78

View Full Product Details