The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006379.3 | NP_006370.1 | 11 - 12 | 1698 | 76 |
XM_005250113.2 | XP_005250170.1 | 12 - 13 | 1619 | 76 | |
GenBank mRNA | AB000220.1 | - | 11 - 12 | 1698 | 76 |
AK304294.1 | - | 11 - 12 | 1269 | 76 | |
AK312478.1 | - | 11 - 12 | 1409 | 76 | |
BC030690.1 | - | 11 - 12 | 1501 | 76 |