The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024957.1 | NP_001020128.1 | 4 - 5 | 506 | 88 |
NM_015399.3 | NP_056214.1 | 4 - 5 | 506 | 88 | |
GenBank mRNA | AF159141.1 | - | 4 - 5 | 480 | 88 |
AI797564.1 | - | 2 - 3 | 88 | ||
AK310991.1 | - | 4 - 5 | 482 | 88 | |
AK313773.1 | - | 4 - 5 | 589 | 88 | |
AL050008.1 | - | 2 - 3 | 215 | 88 | |
BC009834.2 | - | 4 - 5 | 462 | 88 | |
CR457173.1 | - | 3 - 4 | 359 | 88 |