Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENPEP
Entrez Gene ID
2028
Gene Name
glutamyl aminopeptidase
Gene Aliases
APA, CD249, gp160
Chromosome Location
Chr.4: 110476073 - 110563337 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001977.3 NP_001968.3 10 - 11 2062 148
XM_017007877.1 XP_016863366.1 7 - 8 740 148
GenBank mRNA AK313412.1 - 11 - 12 1724 148
AM888273.1 - 10 - 11 2062 148
AM888274.1 - 10 - 11 2062 148
AM888275.1 - 10 - 11 2062 148
AM888276.1 - 10 - 11 2062 148
AM888277.1 - 10 - 11 2062 148
AM888278.1 - 10 - 11 2062 148
AM888279.1 - 10 - 11 2062 148
AM888280.1 - 10 - 11 2062 148
AM888281.1 - 10 - 11 2062 148
AM888282.1 - 10 - 11 2062 148
AM888283.1 - 10 - 11 2062 148
BC094770.1 - 11 - 12 2043 148
BX641012.1 - 7 - 8 1229 148
L12468.1 - 11 - 12 1966 148
L14721.1 - 11 - 12 1803 148

View Full Product Details