The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001977.3 | NP_001968.3 | 17 - 18 | 2843 | 70 |
XM_017007877.1 | XP_016863366.1 | 14 - 15 | 1521 | 70 | |
GenBank mRNA | AK313412.1 | - | 18 - 19 | 2505 | 70 |
AM888273.1 | - | 17 - 18 | 2843 | 70 | |
AM888274.1 | - | 17 - 18 | 2843 | 70 | |
AM888275.1 | - | 17 - 18 | 2843 | 70 | |
AM888276.1 | - | 17 - 18 | 2843 | 70 | |
AM888277.1 | - | 17 - 18 | 2843 | 70 | |
AM888278.1 | - | 17 - 18 | 2843 | 70 | |
AM888279.1 | - | 17 - 18 | 2843 | 70 | |
AM888280.1 | - | 17 - 18 | 2843 | 70 | |
AM888281.1 | - | 17 - 18 | 2843 | 70 | |
AM888282.1 | - | 17 - 18 | 2843 | 70 | |
AM888283.1 | - | 17 - 18 | 2843 | 70 | |
BC094770.1 | - | 18 - 19 | 2824 | 70 | |
BX641012.1 | - | 14 - 15 | 2010 | 70 | |
L12468.1 | - | 18 - 19 | 2747 | 70 | |
L14721.1 | - | 18 - 19 | 2584 | 70 |