The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039848.2 | NP_001034937.1 | 5 - 6 | 694 | 61 |
NM_002085.4 | NP_002076.2 | 5 - 6 | 616 | 61 | |
GenBank mRNA | AI274351.1 | - | 2 - 3 | 61 | |
AK225859.1 | - | 5 - 6 | 535 | 61 | |
BC010157.2 | - | 5 - 6 | 444 | 61 | |
BC011836.2 | - | 5 - 6 | 537 | 61 | |
BC021567.1 | - | 5 - 6 | 530 | 61 | |
BC022071.1 | - | 5 - 6 | 553 | 61 | |
BC032695.1 | - | 5 - 6 | 537 | 61 | |
BC039849.1 | - | 5 - 6 | 605 | 61 | |
BC046163.1 | - | 5 - 6 | 512 | 61 | |
BC058329.1 | - | 5 - 6 | 533 | 61 | |
BI561548.1 | - | 5 - 6 | 612 | 61 | |
BQ648717.1 | - | 5 - 6 | 481 | 61 | |
CF127510.1 | - | 5 - 6 | 628 | 61 | |
X71973.1 | - | 5 - 6 | 578 | 61 |