The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164716.1 | NP_001158188.1 | 9 - 10 | 1954 | 85 |
GenBank mRNA | AF066859.1 | - | 11 - 12 | 1401 | 85 |
AK056607.1 | - | 9 - 10 | 1213 | 85 | |
AK299376.1 | - | 7 - 8 | 741 | 85 | |
AK299461.1 | - | 11 - 12 | 1420 | 85 | |
AK314474.1 | - | 11 - 12 | 1405 | 85 | |
BC126392.1 | - | 11 - 12 | 1800 | 85 | |
BC130514.1 | - | 11 - 12 | 1800 | 85 |