Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELF2
Entrez Gene ID
10659
Gene Name
CUGBP, Elav-like family member 2
Gene Aliases
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location
Chr.10: 10462515 - 11336675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025076.2 NP_001020247.1 13 - 14 1826 69
NM_001083591.1 NP_001077060.1 13 - 14 1820 69
NM_001326320.1 NP_001313249.1 16 - 17 1779 69
NM_001326323.1 NP_001313252.1 17 - 18 1821 69
NM_001326324.1 NP_001313253.1 14 - 15 1662 69
NM_001326325.1 NP_001313254.1 15 - 16 1738 69
NM_001326326.1 NP_001313255.1 14 - 15 1663 69
NM_001326327.1 NP_001313256.1 14 - 15 1681 69
NM_001326328.1 NP_001313257.1 15 - 16 1775 69
NM_001326331.1 NP_001313260.1 13 - 14 1867 69
NM_001326332.1 NP_001313261.1 13 - 14 1849 69
NM_001326335.1 NP_001313264.1 13 - 14 1861 69
NM_001326337.1 NP_001313266.1 12 - 13 1681 69
NM_001326341.1 NP_001313270.1 13 - 14 1761 69
NM_001326344.1 NP_001313273.1 13 - 14 1551 69
NM_001326345.1 NP_001313274.1 13 - 14 1647 69
NM_001326346.1 NP_001313275.1 11 - 12 1794 69
NM_001326347.1 NP_001313276.1 13 - 14 1971 69
NM_001326348.1 NP_001313277.1 13 - 14 1957 69
NM_006561.3 NP_006552.3 13 - 14 1772 69
XM_005252354.4 XP_005252411.1 13 - 14 1862 69
XM_005252358.3 XP_005252415.1 11 - 12 1804 69
XM_006717373.2 XP_006717436.1 12 - 13 1902 69
XM_011519294.2 XP_011517596.1 13 - 14 1844 69
XM_017015542.1 XP_016871031.1 12 - 13 1550 69
XM_017015546.1 XP_016871035.1 13 - 14 1622 69
XM_017015549.1 XP_016871038.1 13 - 14 1610 69
XM_017015551.1 XP_016871040.1 13 - 14 1856 69
XM_017015554.1 XP_016871043.1 13 - 14 1967 69
XM_017015569.1 XP_016871058.1 12 - 13 1899 69
XM_017015572.1 XP_016871061.1 12 - 13 1588 69
GenBank mRNA AF036956.1 - 13 - 14 1742 69
AF090694.1 - 13 - 14 1610 69
AF432906.1 - 13 - 14 1672 69
AK293164.1 - 13 - 14 1713 69
AK297313.1 - 11 - 12 1585 69
AK299548.1 - 13 - 14 1602 69
CR976278.1 - 3 - 4 413 69
U69546.1 - 13 - 14 1559 69

View Full Product Details