The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006019.3 | NP_006010.2 | 16 - 17 | 2146 | 119 |
NM_006053.3 | NP_006044.1 | 11 - 12 | 1905 | 119 | |
XM_005273709.3 | XP_005273766.1 | 17 - 18 | 2305 | 119 | |
XM_011544726.2 | XP_011543028.1 | 16 - 17 | 2045 | 119 | |
XM_017017089.1 | XP_016872578.1 | 16 - 17 | 2141 | 119 | |
XM_017017090.1 | XP_016872579.1 | 15 - 16 | 2484 | 119 | |
GenBank mRNA | AF025374.1 | - | 11 - 12 | 1905 | 119 |
AY548970.1 | - | 1 - 2 | 645 | 119 | |
BC018133.1 | - | 16 - 17 | 2125 | 119 | |
BC032465.1 | - | 16 - 17 | 2118 | 119 | |
U45285.1 | - | 16 - 17 | 2071 | 119 |