Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RC3H2
Entrez Gene ID
54542
Gene Name
ring finger and CCCH-type domains 2
Gene Aliases
MNAB, RNF164
Chromosome Location
Chr.9: 122849453 - 122905283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100588.1 NP_001094058.1 14 - 15 2842 106
NM_018835.2 NP_061323.2 14 - 15 2842 106
GenBank mRNA AF255303.1 - 13 - 14 3202 106
AF255304.1 - 2 - 3 286 106
AK000308.1 - 12 - 13 2276 106
AK000720.1 - 1 - 2 69 106
AK027042.1 - 5 - 6 1079 106
AK226010.1 - 12 - 13 2276 106
AL833177.1 - 15 - 16 2699 106
AL834431.1 - 8 - 9 1593 106

View Full Product Details