Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSSC4
Entrez Gene ID
10078
Gene Name
tumor suppressing subtransferable candidate 4
Gene Aliases
-
Chromosome Location
Chr.11: 2400409 - 2403878 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297658.1 NP_001284587.1 1 - 2 313 87
NM_001297660.1 NP_001284589.1 1 - 2 313 87
GenBank mRNA BX384913.2 - 1 - 2 42 87

View Full Product Details