Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSSC4
Entrez Gene ID
10078
Gene Name
tumor suppressing subtransferable candidate 4
Gene Aliases
-
Chromosome Location
Chr.11: 2400409 - 2403878 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297661.1 NP_001284590.1 2 - 3 469 150
GenBank mRNA BC050616.1 - 2 - 3 469 150

View Full Product Details