Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRFIP2
Entrez Gene ID
9209
Gene Name
LRR binding FLII interacting protein 2
Gene Aliases
HUFI-2
Chromosome Location
Chr.3: 37052626 - 37176817 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006309.3 NP_006300.1 14 - 15 905 68
XM_005265539.2 XP_005265596.1 15 - 16 1905 68
XM_005265540.1 XP_005265597.1 16 - 17 1188 68
XM_005265550.1 XP_005265607.1 7 - 8 741 68
XM_006713385.1 XP_006713448.1 15 - 16 1170 68
XM_006713386.1 XP_006713449.1 14 - 15 1131 68
XM_006713387.1 XP_006713450.1 14 - 15 1128 68
XM_006713388.1 XP_006713451.1 14 - 15 1125 68
XM_006713389.1 XP_006713452.1 14 - 15 1122 68
XM_006713390.1 XP_006713453.1 15 - 16 1165 68
XM_006713392.1 XP_006713455.1 15 - 16 1173 68
XM_006713394.1 XP_006713457.1 13 - 14 1080 68
XM_006713395.1 XP_006713458.1 15 - 16 1173 68
XM_006713396.1 XP_006713459.1 13 - 14 1065 68
XM_011534217.1 XP_011532519.1 15 - 16 1173 68
XM_011534218.1 XP_011532520.1 16 - 17 1216 68
XM_011534219.1 XP_011532521.1 16 - 17 1284 68
XM_011534221.1 XP_011532523.1 13 - 14 1065 68
XM_011534222.1 XP_011532524.1 15 - 16 1173 68
XM_017007465.1 XP_016862954.1 12 - 13 1020 68
XM_017007467.1 XP_016862956.1 15 - 16 1173 68
XM_017007468.1 XP_016862957.1 10 - 11 912 68
XM_017007469.1 XP_016862958.1 15 - 16 1173 68
XM_017007470.1 XP_016862959.1 12 - 13 1035 68
GenBank mRNA AF115509.1 - 14 - 15 864 68

View Full Product Details