Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRFIP2
Entrez Gene ID
9209
Gene Name
LRR binding FLII interacting protein 2
Gene Aliases
HUFI-2
Chromosome Location
Chr.3: 37052626 - 37176817 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006309.3 NP_006300.1 7 - 8 483 135
XM_005265539.2 XP_005265596.1 8 - 9 1483 135
XM_005265540.1 XP_005265597.1 9 - 10 766 135
XM_006713385.1 XP_006713448.1 8 - 9 751 135
XM_006713387.1 XP_006713450.1 8 - 9 751 135
XM_006713388.1 XP_006713451.1 8 - 9 751 135
XM_006713389.1 XP_006713452.1 7 - 8 700 135
XM_006713390.1 XP_006713453.1 8 - 9 743 135
XM_006713392.1 XP_006713455.1 8 - 9 751 135
XM_006713393.1 XP_006713456.1 8 - 9 751 135
XM_006713395.1 XP_006713458.1 8 - 9 751 135
XM_006713396.1 XP_006713459.1 6 - 7 643 135
XM_006713397.1 XP_006713460.1 8 - 9 751 135
XM_011534217.1 XP_011532519.1 8 - 9 751 135
XM_011534218.1 XP_011532520.1 9 - 10 794 135
XM_011534219.1 XP_011532521.1 9 - 10 862 135
XM_011534222.1 XP_011532524.1 8 - 9 751 135
XM_017007466.1 XP_016862955.1 8 - 9 751 135
XM_017007467.1 XP_016862956.1 8 - 9 751 135
XM_017007469.1 XP_016862958.1 8 - 9 751 135
GenBank mRNA AF115509.1 - 7 - 8 442 135

View Full Product Details