The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021185.4 | NP_067008.3 | 4 - 5 | 563 | 79 |
XM_005259114.4 | XP_005259171.1 | 4 - 5 | 659 | 79 | |
XM_005259115.4 | XP_005259172.1 | 4 - 5 | 582 | 79 | |
XM_005259116.2 | XP_005259173.1 | 4 - 5 | 563 | 79 | |
XM_006723308.3 | XP_006723371.1 | 4 - 5 | 1179 | 79 | |
XM_006723310.2 | XP_006723373.1 | 4 - 5 | 563 | 79 | |
XM_006723311.2 | XP_006723374.1 | 4 - 5 | 584 | 79 | |
XM_011527174.2 | XP_011525476.1 | 4 - 5 | 573 | 79 | |
XM_011527175.2 | XP_011525477.1 | 4 - 5 | 479 | 79 | |
XM_011527176.2 | XP_011525478.1 | 4 - 5 | 563 | 79 | |
XM_011527178.2 | XP_011525480.1 | 5 - 6 | 925 | 79 | |
XM_011527179.1 | XP_011525481.1 | 4 - 5 | 563 | 79 | |
XM_011527182.2 | XP_011525484.1 | 4 - 5 | 479 | 79 | |
XM_017027053.1 | XP_016882542.1 | 4 - 5 | 563 | 79 | |
XM_017027054.1 | XP_016882543.1 | 2 - 3 | 176 | 79 | |
GenBank mRNA | AK128220.1 | - | 4 - 5 | 479 | 79 |
AK310460.1 | - | 1 - 2 | 65 | 79 | |
AL133023.1 | - | 4 - 5 | 1179 | 79 | |
BC094785.1 | - | 5 - 6 | 655 | 79 |