Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBPL6
Entrez Gene ID
114880
Gene Name
oxysterol binding protein like 6
Gene Aliases
ORP6
Chromosome Location
Chr.2: 178194479 - 178399433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201480.1 NP_001188409.1 9 - 10 1341 76
NM_001201481.1 NP_001188410.1 9 - 10 1341 76
NM_001201482.1 NP_001188411.1 9 - 10 1341 76
NM_032523.3 NP_115912.1 9 - 10 1341 76
NM_145739.2 NP_665682.1 7 - 8 854 76
XM_011510539.1 XP_011508841.1 8 - 9 1244 76
XM_017003265.1 XP_016858754.1 10 - 11 1413 76
XM_017003266.1 XP_016858755.1 10 - 11 1413 76
XM_017003267.1 XP_016858756.1 8 - 9 1108 76
XM_017003268.1 XP_016858757.1 9 - 10 1337 76
XM_017003269.1 XP_016858758.1 8 - 9 1108 76
XM_017003270.1 XP_016858759.1 10 - 11 1495 76
XM_017003271.1 XP_016858760.1 8 - 9 1108 76
XM_017003272.1 XP_016858761.1 8 - 9 1244 76
XM_017003273.1 XP_016858762.1 8 - 9 1244 76
XM_017003274.1 XP_016858763.1 9 - 10 1337 76
XM_017003275.1 XP_016858764.1 10 - 11 1431 76
XM_017003276.1 XP_016858765.1 8 - 9 1244 76
XM_017003277.1 XP_016858766.1 8 - 9 1244 76
GenBank mRNA AB208898.1 - 9 - 10 1341 76
AF323728.1 - 9 - 10 1175 76
AF392448.1 - 9 - 10 1175 76
AF462443.1 - 8 - 9 1151 76
AK027600.1 - 2 - 3 216 76
AK093902.1 - 7 - 8 856 76
AK300389.1 - 9 - 10 1299 76
BC021248.2 - 1 - 2 153 76
BC052259.1 - 9 - 10 1322 76

View Full Product Details