Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CATSPERG
- Entrez Gene ID
-
57828
- Gene Name
- cation channel sperm associated auxiliary subunit gamma
- Gene Aliases
-
C19orf15
- Chromosome Location
-
Chr.19: 38335563 - 38370949 on Build GRCh38
View Full Product Details