The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134369.2 | NP_001127841.1 | 4 - 5 | 606 | 98 |
NM_017724.2 | NP_060194.1 | 4 - 5 | 606 | 98 | |
XM_005265553.1 | XP_005265610.1 | 4 - 5 | 573 | 98 | |
XM_005265554.2 | XP_005265611.1 | 4 - 5 | 1305 | 98 | |
XM_005265556.1 | XP_005265613.1 | 4 - 5 | 573 | 98 | |
XM_017007476.1 | XP_016862965.1 | 5 - 6 | 616 | 98 | |
XM_017007478.1 | XP_016862967.1 | 4 - 5 | 573 | 98 | |
XM_017007480.1 | XP_016862969.1 | 4 - 5 | 817 | 98 | |
XM_017007481.1 | XP_016862970.1 | 4 - 5 | 573 | 98 | |
XM_017007482.1 | XP_016862971.1 | 4 - 5 | 573 | 98 | |
XM_017007483.1 | XP_016862972.1 | 4 - 5 | 1305 | 98 | |
XM_017007484.1 | XP_016862973.1 | 5 - 6 | 616 | 98 | |
GenBank mRNA | AK000255.1 | - | 3 - 4 | 291 | 98 |
AK291514.1 | - | 4 - 5 | 583 | 98 | |
BC053668.1 | - | 4 - 5 | 563 | 98 | |
DB033703.1 | - | 4 - 5 | 359 | 98 |