The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134369.2 | NP_001127841.1 | 9 - 10 | 1098 | 88 |
NM_017724.2 | NP_060194.1 | 8 - 9 | 1026 | 88 | |
XM_006713404.1 | XP_006713467.1 | 9 - 10 | 1038 | 88 | |
XM_011534222.1 | XP_011532524.1 | 21 - 22 | 1761 | 88 | |
XM_017007469.1 | XP_016862958.1 | 20 - 21 | 1689 | 88 | |
XM_017007479.1 | XP_016862968.1 | 11 - 12 | 1167 | 88 | |
XM_017007483.1 | XP_016862972.1 | 9 - 10 | 1797 | 88 | |
XM_017007484.1 | XP_016862973.1 | 10 - 11 | 1108 | 88 | |
GenBank mRNA | AK000255.1 | - | 7 - 8 | 711 | 88 |
AK024692.1 | - | 1 - 2 | 44 | 88 | |
AK291514.1 | - | 9 - 10 | 1075 | 88 | |
BC053668.1 | - | 8 - 9 | 983 | 88 | |
BX648794.1 | - | 5 - 6 | 477 | 88 |