The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201482.1 | NP_001188411.1 | 11 - 12 | 1530 | 90 |
NM_032523.3 | NP_115912.1 | 11 - 12 | 1530 | 90 | |
XM_017003271.1 | XP_016858760.1 | 10 - 11 | 1297 | 90 | |
XM_017003276.1 | XP_016858765.1 | 10 - 11 | 1433 | 90 | |
GenBank mRNA | AF323728.1 | - | 11 - 12 | 1364 | 90 |
AF392448.1 | - | 11 - 12 | 1364 | 90 | |
AF462443.1 | - | 10 - 11 | 1340 | 90 | |
AK027600.1 | - | 4 - 5 | 405 | 90 | |
BC021248.2 | - | 3 - 4 | 342 | 90 |