The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318245.1 | NP_001305174.1 | 26 - 27 | 4694 | 105 |
NM_032608.6 | NP_115997.5 | 26 - 27 | 4691 | 105 | |
XM_011530458.2 | XP_011528760.1 | 27 - 28 | 4830 | 105 | |
XM_011530459.2 | XP_011528761.1 | 27 - 28 | 4801 | 105 | |
XM_011530460.2 | XP_011528762.1 | 26 - 27 | 4674 | 105 | |
XM_011530461.2 | XP_011528763.1 | 27 - 28 | 4830 | 105 | |
XM_011530464.2 | XP_011528766.1 | 27 - 28 | 4827 | 105 | |
XM_011530465.2 | XP_011528767.1 | 26 - 27 | 4587 | 105 | |
XM_011530466.2 | XP_011528768.1 | 27 - 28 | 4831 | 105 | |
XM_017029012.1 | XP_016884501.1 | 27 - 28 | 4830 | 105 | |
XM_017029013.1 | XP_016884502.1 | 27 - 28 | 4878 | 105 | |
XM_017029014.1 | XP_016884503.1 | 27 - 28 | 4830 | 105 | |
XM_017029015.1 | XP_016884504.1 | 26 - 27 | 4701 | 105 | |
XM_017029016.1 | XP_016884505.1 | 15 - 16 | 1954 | 105 | |
XM_017029017.1 | XP_016884506.1 | 25 - 26 | 4555 | 105 | |
GenBank mRNA | AB042648.2 | - | 22 - 23 | 3003 | 105 |
AB075376.1 | - | 26 - 27 | 4691 | 105 | |
AJ310931.2 | - | 26 - 27 | 4682 | 105 | |
AY077700.1 | - | 26 - 27 | 4604 | 105 | |
BC144596.1 | - | 24 - 25 | 4346 | 105 | |
BC144597.1 | - | 25 - 26 | 4491 | 105 | |
BC150626.1 | - | 25 - 26 | 4491 | 105 |