Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO18B
Entrez Gene ID
84700
Gene Name
myosin XVIIIB
Gene Aliases
KFS4
Chromosome Location
Chr.22: 25742133 - 26063847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318245.1 NP_001305174.1 36 - 37 6014 68
NM_032608.6 NP_115997.5 36 - 37 6011 68
XM_011530458.2 XP_011528760.1 37 - 38 6150 68
XM_011530459.2 XP_011528761.1 37 - 38 6121 68
XM_011530460.2 XP_011528762.1 36 - 37 5994 68
XM_011530461.2 XP_011528763.1 37 - 38 6150 68
XM_011530464.2 XP_011528766.1 37 - 38 6147 68
XM_011530465.2 XP_011528767.1 36 - 37 5907 68
XM_011530466.2 XP_011528768.1 37 - 38 6151 68
XM_017029012.1 XP_016884501.1 37 - 38 6150 68
XM_017029013.1 XP_016884502.1 37 - 38 6198 68
XM_017029014.1 XP_016884503.1 37 - 38 6147 68
XM_017029015.1 XP_016884504.1 36 - 37 6018 68
XM_017029016.1 XP_016884505.1 25 - 26 3274 68
XM_017029017.1 XP_016884506.1 35 - 36 5875 68
GenBank mRNA AB042648.2 - 32 - 33 4323 68
AB075376.1 - 36 - 37 6011 68
AJ310931.2 - 36 - 37 6002 68
AY077700.1 - 36 - 37 5924 68
BC144596.1 - 34 - 35 5666 68
BC144597.1 - 35 - 36 5808 68
BC150626.1 - 35 - 36 5808 68
CR749556.1 - 1 - 2 68

View Full Product Details